2-Propynylurea
Supplier: Apollo Scientific
Warning
Synonyms: 1-(Prop-2-yn-1-yl)urea, Propargylurea
OR23964-1G OR23964-5G
APOSOR23964-1GEA 77 EUR
APOSOR23964-1G APOSOR23964-5G
2-Propynylurea
2-Propynylurea
Formula: C₄H₆N₂O
Melting Pt: 130…131 °C
Storage Temperature: Ambient
MDL Number: MFCD00052817
CAS Number: 5221-62-5