3-Chloro-4-methylphenyl isocyanate 97%
Supplier: Apollo Scientific
Danger
Synonyms: 2-Chloro-4-isocyanato-1-methylbenzene
OR23984-100G OR23984-25G OR23984-5G
APOSOR23984-100GEA 331 EUR
APOSOR23984-100G APOSOR23984-25G APOSOR23984-5G
3-Chloro-4-methylphenyl isocyanate 97%
3-Chloro-4-methylphenyl isocyanate
  • May form small amount of sediment.
Formula: C₈H₆ClNO
MW: 167.59 g/mol
Boiling Pt: 107 °C/3mmHg
Melting Pt: 20…24 °C
Density: 1.224 g/cm³ (25 °C)
Flash Pt: 109 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00013858
CAS Number: 28479-22-3
EINECS: 249-050-7
UN: 2236
ADR: 6.1 II