4-Propargylthiomorpholine-1,1-dioxide
Supplier: Apollo Scientific
Danger
Synonyms: 4-(2-Propynyl)thiomorpholine 1,1-Dioxide, 4-(Prop-2-yn-1-yl)thiomorpholine 1,1-dioxide
OR24044-1G OR24044-250MG OR24044-5G
APOSOR24044-1GEA 60.9 EUR
APOSOR24044-1G APOSOR24044-250MG APOSOR24044-5G
4-Propargylthiomorpholine-1,1-dioxide
4-Propargylthiomorpholine-1,1-dioxide
Formula: C₇H₁₁NO₂S
MW: 173.23 g/mol
Melting Pt: 114…116 °C
Storage Temperature: Ambient
MDL Number: MFCD00085157
CAS Number: 10442-03-2
UN: 3259
ADR: 8 III