3,5-Dioxocyclohexanecarboxylic acid 97%
Supplier: Apollo Scientific
Warning
OR2410-1G OR2410-25G OR2410-5G
APOSOR2410-1GEA 46.4 EUR
APOSOR2410-1G APOSOR2410-25G APOSOR2410-5G
3,5-Dioxocyclohexanecarboxylic acid 97%
3,5-Dioxocyclohexanecarboxylic acid
Formula: C₇H₈O₄
MW: 156.1360 g/mol
Melting Pt: 180…184 °C
Storage Temperature: Ambient
MDL Number: MFCD09836162
CAS Number: 42858-60-6