2-Hydrazino-1,4,5,6-tetrahydropyrimidinehydrobromide
Supplier: Apollo Scientific
Warning
OR24100-1G OR24100-250MG OR24100-500MG OR24100-2.5G
APOSOR24100-1GEA 353 EUR
APOSOR24100-1G APOSOR24100-250MG APOSOR24100-500MG APOSOR24100-2.5G
2-Hydrazino-1,4,5,6-tetrahydropyrimidinehydrobromide
2-Hydrazino-1,4,5,6-tetrahydropyrimidinehydrobromide
Formula: C₄H₁₁BrN₄
MW: 195.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00053056
CAS Number: 197234-18-7