2,5-Dichlorobenzoic acid
Supplier: Apollo Scientific
Warning
OR24122-100G OR24122-25G OR24122-500G
APOSOR24122-100GEA 36.4 EUR
APOSOR24122-100G APOSOR24122-25G APOSOR24122-500G
2,5-Dichlorobenzoic acid
2,5-Dichlorobenzoic acid
Formula: C₇H₄Cl₂O₂
MW: 191.01 g/mol
Boiling Pt: 300…302 °C
Melting Pt: 151…154 °C
Storage Temperature: Ambient
MDL Number: MFCD00002416
CAS Number: 50-79-3
EINECS: 200-065-7