2-Bromo-p-cresol 97%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Bromo-4-methylphenol, 1-Bromo-2-hydroxy-5-methylbenzene
OR2424-100G OR2424-25G OR2424-500G
APOSOR2424-100GEA 66.7 EUR
APOSOR2424-100G APOSOR2424-25G APOSOR2424-500G
2-Bromo-p-cresol 97%
2-Bromo-p-cresol
Formula: C₇H₇BrO
MW: 187.04 g/mol
Boiling Pt: 213…214 °C
Melting Pt: 16 °C
Density: 1.547 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00002151
CAS Number: 6627-55-0
EINECS: 229-595-7