3-Amino-1-methyl-1H-pyrazole
Supplier: Apollo Scientific
Warning
Synonyms: 1-Methyl-1H-pyrazol-3-amine, 3-Amino-1-methylpyrazole, 1-Methyl-1H-pyrazol-3-ylamine, 3-Amino-1-methyl-1H-pyrazole
OR24245-25G OR24245-5G OR24245-100G
APOSOR24245-25GEA 55.1 EUR
APOSOR24245-25G APOSOR24245-5G APOSOR24245-100G
3-Amino-1-methyl-1H-pyrazole
3-Amino-1-methyl-1H-pyrazole
Formula: C₄H₇N₃
MW: 97.12 g/mol
Boiling Pt: 90…92 °C/1mmHg
Density: 1.12 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00466340
CAS Number: 1904-31-0