N,N-Dimethylcyanoacetamide

Supplier: Apollo Scientific
Warning

OR24266-5G OR24266-25G
APOSOR24266-5GEA 38.7 EUR
APOSOR24266-5G APOSOR24266-25G
N,N-Dimethylcyanoacetamide
N,N-Dimethylcyanoacetamide
Formula: C₅H₈N₂O
Melting Pt: 59…66 °C
Storage Temperature: Ambient
MDL Number: MFCD00051623
CAS Number: 7391-40-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR