6-Bromo-1,4-benzodioxane 98%
Supplier: Apollo Scientific
Warning
OR2430-100G OR2430-25G OR2430-5G
APOSOR2430-100GEA 236 EUR
APOSOR2430-100G APOSOR2430-25G APOSOR2430-5G
6-Bromo-1,4-benzodioxane 98%
6-Bromo-1,4-benzodioxane
Formula: C₈H₇BrO₂
Boiling Pt: 259…260 °C
Density: 1.598 g/cm³ (25 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00040750
CAS Number: 52287-51-1
EINECS: 257-817-2