4-Chloro-5-methylthieno[2,3-d]pyrimidine
Supplier: Apollo Scientific
|
Warning
|
OR24334-1G
OR24334-5G
APOSOR24334-1GEA
50.9
EUR
APOSOR24334-1G
APOSOR24334-5G
4-Chloro-5-methylthieno[2,3-d]pyrimidine
4-Chloro-5-methylthieno[2,3-d]pyrimidine
Formula:
C₇H₅ClN₂S Melting Pt: 138…140 °C Storage Temperature: Ambient |
MDL Number:
MFCD00205201 CAS Number: 43088-67-1 |