Imipenem monohydrate

Supplier: Apollo Scientific
Warning

APOSOR2453-1GEA 374 EUR
APOSOR2453-1G
Imipenem monohydrate
Imipenem monohydrate
Formula: C₁₂H₁₇N₃O₄S
MW: 317,36 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09753321
CAS Number: 74431-23-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR