2-Nitro-1-naphthol 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1-Hydroxy-2-nitronaphthalene
OR24530-1G OR24530-5G
APOSOR24530-1GEA 21.9 EUR
APOSOR24530-1G APOSOR24530-5G
2-Nitro-1-naphthol 98%
2-Nitro-1-naphthol
Formula: C₁₀H₇NO₃
MW: 189.17 g/mol
Melting Pt: 123…125 °C
Storage Temperature: Ambient
MDL Number: MFCD00003956
CAS Number: 607-24-9