8-Hydroxyjulolidine 97%
Supplier: Apollo Scientific
Warning
Synonyms: 1,2,3,5,6,7-Hexahydropyrido[3,2,1-ij]quinolin-8-ol, 2,3,6,7-Tetrahydro-1H,5H-benzo[ij]quinolizin-8-ol
OR24664-5G OR24664-1G
APOSOR24664-5GEA 212 EUR
APOSOR24664-5G APOSOR24664-1G
8-Hydroxyjulolidine 97%
8-Hydroxyjulolidine
Formula: C₁₂H₁₅NO
MW: 189.26 g/mol
Melting Pt: 132…134 °C
Storage Temperature: Ambient
MDL Number: MFCD00006918
CAS Number: 41175-50-2
EINECS: 255-247-9