4-Chlorothieno[2,3-d]pyrimidine
Supplier: Apollo Scientific
|
Warning
|
OR2473-5G
OR2473-1G
OR2473-10G
APOSOR2473-5GEA
102
EUR
APOSOR2473-5G
APOSOR2473-1G
APOSOR2473-10G
4-Chlorothieno[2,3-d]pyrimidine
4-Chlorothieno[2,3-d]pyrimidine
Formula:
C₆H₃ClN₂S MW: 170.6200 g/mol Melting Pt: 105 °C Storage Temperature: Ambient |
MDL Number:
MFCD01312373 CAS Number: 14080-59-2 |