1-Bromo-5-chloropentane
Supplier: Apollo Scientific
Warning
OR24918-100G OR24918-25G
APOSOR24918-100GEA 24.8 EUR
APOSOR24918-100G APOSOR24918-25G
1-Bromo-5-chloropentane
1-Bromo-5-chloropentane
Formula: BrH₂C(CH₂)₃CH₂Cl
MW: 185.49 g/mol
Boiling Pt: 210…212 °C
Density: 1.408 g/cm³ (25 °C)
Flash Pt: 95 °C
Storage Temperature: Ambient
MDL Number: MFCD00001016
CAS Number: 54512-75-3
EINECS: 259-193-7