1-Methylisoquinoline
Supplier: Apollo Scientific
Warning
OR24943-25G OR24943-5G OR24943-1G OR24943-250MG OR24943-10G
APOSOR24943-25GEA 522 EUR
APOSOR24943-25G APOSOR24943-5G APOSOR24943-1G APOSOR24943-250MG APOSOR24943-10G
1-Methylisoquinoline
1-Methylisoquinoline
Formula: C₁₀H₉N
MW: 143.19 g/mol
Boiling Pt: 126…128 °C/16mmHg
Melting Pt: 44540 °C
Density: 1.078 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00006902
CAS Number: 1721-93-3
EINECS: 217-017-6