1,3-Dimethyl-1H-pyrazole 96+%
Supplier: Apollo Scientific
Warning
Synonyms: 1,3-Dimethylpyrazole
OR2502-100G
APOSOR2502-100GEA 95.7 EUR
APOSOR2502-100G
1,3-Dimethyl-1H-pyrazole 96+%
1,3-Dimethyl-1H-pyrazole
Formula: C₅H₈N₂
MW: 96.1304 g/mol
Boiling Pt: 135…139 °C
Density: 0.96 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00036665
CAS Number: 694-48-4