4-Methoxythiophene-3-carboxylic acid
Supplier: Apollo Scientific
Warning
OR25041-1G OR25041-5G OR25041-250MG
APOSOR25041-1GEA 65.4 EUR
APOSOR25041-1G APOSOR25041-5G APOSOR25041-250MG
4-Methoxythiophene-3-carboxylic acid
Formula: C₆H₆O₃S
Melting Pt: 101…105 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00067997
CAS Number: 71050-40-3