Methyl p-tolyl sulphoxide
Supplier: Apollo Scientific
Warning
Synonyms: 1-Methyl-4-(methylsulphinyl)benzene, Methyl 4-Tolyl sulphoxide
OR25242-1G OR25242-5G
APOSOR25242-1GEA 43.5 EUR
APOSOR25242-1G APOSOR25242-5G
Methyl p-tolyl sulphoxide
Methyl p-tolyl sulphoxide
Formula: C₈H₁₀OS
MW: 154.227 g/mol
Boiling Pt: 113…114 °C/2mmHg
Melting Pt: 44…46 °C
Flash Pt: >113 °C
Storage Temperature: Ambient
MDL Number: MFCD00075175
CAS Number: 934-72-5