2-Chlorobenzylideneacetone
Supplier: Apollo Scientific
Synonyms:
4-(2-Chlorophenyl)but-3-en-2-one
OR25346-1G
OR25346-5G
APOSOR25346-1GEA
0
EUR
APOSOR25346-1G
APOSOR25346-5G
2-Chlorobenzylideneacetone
2-Chlorobenzylideneacetone
Formula:
C₁₀H₉ClO Storage Temperature: Ambient |
CAS Number:
20766-37-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...