2-Bromo-1-methyl-1H-imidazole-4,5-dicarbonitrile
Supplier: Apollo Scientific
OR25565-1G OR25565-250MG OR25565-500MG OR25565-5G
APOSOR25565-1GEA 258 EUR
APOSOR25565-1G APOSOR25565-250MG APOSOR25565-500MG APOSOR25565-5G
2-Bromo-1-methyl-1H-imidazole-4,5-dicarbonitrile
2-Bromo-1-methyl-1H-imidazole-4,5-dicarbonitrile
Formula: C₆H₃BrN₄
Storage Temperature: Ambient
MDL Number: MFCD00174289
CAS Number: 115905-43-6