(S)-(-)-1-(1-Naphthyl)ethylamine 97%
Supplier: Apollo Scientific
Danger
Synonyms: (S)-(-)-alpha-(1-Naphthyl)ethylamine
OR2560-25G OR2560-5G OR2560-100G
APOSOR2560-25GEA 55.1 EUR
APOSOR2560-25G APOSOR2560-5G APOSOR2560-100G
(S)-(-)-1-(1-Naphthyl)ethylamine 97%
(S)-(-)-1-(1-Naphthyl)ethylamine
Formula: C₁₂H₁₃N
MW: 171.24 g/mol
Boiling Pt: 152…153 °C/11mmHg
Density: 1.06 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00064179
CAS Number: 10420-89-0
UN: 2922
ADR: 8+6.1 III