2-Amino-5-bromo-3-methylpyridine 95%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Amino-5-bromo-3-picoline, 5-Bromo-3-methylpyridin-2-amine, 5-Bromo-3-methyl-2-pyridylamine
OR2561-100G OR2561-25G OR2561-500G
APOSOR2561-100GEA 79.9 EUR
APOSOR2561-100G APOSOR2561-25G APOSOR2561-500G
2-Amino-5-bromo-3-methylpyridine 95%
2-Amino-5-bromo-3-methylpyridine
Formula: C₆H₇BrN₂
MW: 187.04 g/mol
Melting Pt: 88…95 °C
Storage Temperature: Ambient
MDL Number: MFCD00068232
CAS Number: 3430-21-5