α-Chloro-m-xylene 94%
Supplier: Apollo Scientific
Danger
Synonyms: 1-(Chloromethyl)-3-methylbenzene, 3-Methylbenzyl chloride
OR2562-100G OR2562-250G OR2562-1KG
APOSOR2562-100GEA 26.1 EUR
APOSOR2562-100G APOSOR2562-250G APOSOR2562-1KG
α-Chloro-m-xylene 94%
α-Chloro-m-xylene
Formula: C₈H₉Cl
MW: 140.61 g/mol
Boiling Pt: 195…196 °C
Density: 1.064 g/cm³ (25 °C)
Flash Pt: 76 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000909
CAS Number: 620-19-9
EINECS: 210-628-9
UN: 3265
ADR: 8 II
Merck Index: 12,10224