4-Methoxybenzyl chloride
Supplier: Apollo Scientific
Danger
Synonyms: 4-(Chloromethyl)anisole, p-(Chloromethyl)anisole, p-Methoxybenzyl chloride
OR25768-100G OR25768-25G OR25768-5G
APOSOR25768-100GEA 249 EUR
APOSOR25768-100G APOSOR25768-25G APOSOR25768-5G
4-Methoxybenzyl chloride
4-Methoxybenzyl chloride
Formula: C₈H₉ClO
Boiling Pt: 117…118 °C/14mmHg
Melting Pt: –1 °C
Density: 1.15 g/cm³ (25 °C)
Flash Pt: 109 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000915
CAS Number: 824-94-2
EINECS: 212-540-6
UN: 3265
ADR: 8,III