N-(3-Formyl-pyridin-4-yl)-2,2-dimethylpropionamide
Supplier: Apollo Scientific
Warning
Synonyms: 2,2-Dimethyl-N-(3-formylpyridin-4-yl)propanamide, N-(3-Formylpyridin-4-yl)pivalamide
OR25911-250MG OR25911-100MG
APOSOR25911-250MGEA 27.7 EUR
APOSOR25911-250MG APOSOR25911-100MG
N-(3-Formyl-pyridin-4-yl)-2,2-dimethylpropionamide
N-(3-Formyl-pyridin-4-yl)-2,2-dimethylpropionamide
Formula: C₁₁H₁₄N₂O₂
MW: 206.25 g/mol
Melting Pt: 60…63 °C
Storage Temperature: Ambient
MDL Number: MFCD03086208
CAS Number: 86847-71-4