2-Carboxythiophene-5-boronic acid
Supplier: Apollo Scientific
Warning
OR25978-1G OR25978-250MG OR25978-5G
APOSOR25978-1GEA 27.7 EUR
APOSOR25978-1G APOSOR25978-250MG APOSOR25978-5G
2-Carboxythiophene-5-boronic acid
2-Carboxythiophene-5-boronic acid
Formula: C₅H₅BO₄S
MW: 171.9668 g/mol
Melting Pt: 135…136 °C
Storage Temperature: Ambient
MDL Number: MFCD02323211
CAS Number: 465515-31-5