Methyl-5-(chlorosulphonyl)-4-methylthiophene-2-carboxylate
Supplier: Apollo Scientific
Danger
OR25981-1G OR25981-250MG OR25981-500MG OR25981-2.5G OR25981-5G
APOSOR25981-1GEA 366 EUR
APOSOR25981-1G APOSOR25981-250MG APOSOR25981-500MG APOSOR25981-2.5G APOSOR25981-5G
Methyl-5-(chlorosulphonyl)-4-methylthiophene-2-carboxylate
Methyl-5-(chlorosulphonyl)-4-methylthiophene-2-carboxylate
Formula: C₇H₇ClO₄S₂
MW: 254.71 g/mol
Melting Pt: 40…41 °C
Storage Temperature: Ambient
MDL Number: MFCD03407367
CAS Number: 423768-46-1
UN: 3261
ADR: 8 III