Benzo[b]thiophene-1,1-dioxide
Supplier: Apollo Scientific
Danger
Synonyms: Benzothiophene 1,1-Dioxide
OR26005-1G OR26005-5G
APOSOR26005-1GEA 27.7 EUR
APOSOR26005-1G APOSOR26005-5G
Benzo[b]thiophene-1,1-dioxide
Benzo[b]thiophene-1,1-dioxide
Formula: C₈H₆O₂S
MW: 166.20 g/mol
Melting Pt: 137…142 °C
Storage Temperature: Ambient
MDL Number: MFCD00052178
CAS Number: 825-44-5
EINECS: 212-544-8
UN: 2811
ADR: 6.1,III