3-(Bromomethyl)-5-chlorobenzo[b]thiophene
Supplier: Apollo Scientific
Danger
OR26008-1G OR26008-5G
APOSOR26008-1GEA 77 EUR
APOSOR26008-1G APOSOR26008-5G
3-(Bromomethyl)-5-chlorobenzo[b]thiophene
3-(Bromomethyl)-5-chlorobenzo[b]thiophene
Formula: C₉H₆BrClS
MW: 261.5700 g/mol
Melting Pt: 129…131 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00173769
CAS Number: 1198-51-2
UN: 3261
ADR: 8,III