5-Chlorobenzo[b]thiophene-3-acetonitrile
Supplier: Apollo Scientific
Danger
OR26011-1G OR26011-5G
APOSOR26011-1GEA 132 EUR
APOSOR26011-1G APOSOR26011-5G
5-Chlorobenzo[b]thiophene-3-acetonitrile
5-Chlorobenzo[b]thiophene-3-acetonitrile
Formula: C₁₀H₆ClNS
Melting Pt: 133…135 °C
Storage Temperature: Ambient
MDL Number: MFCD00052508
CAS Number: 23799-60-2
UN: 3439
ADR: 6.1,III