Benzo[cd]indol-2(1H)-one 95%
Supplier: Apollo Scientific
Warning
Synonyms: Naphthostyril, Naphtholactam, Benz[cd]indol-2(1H)-one
OR26128-100G OR26128-25G OR26128-5G
APOSOR26128-100GEA 79.9 EUR
APOSOR26128-100G APOSOR26128-25G APOSOR26128-5G
Benzo[cd]indol-2(1H)-one 95%
Benzo[cd]indol-2(1H)-one
Formula: C₁₁H₇NO
MW: 169.18 g/mol
Melting Pt: 173…178 °C
Storage Temperature: Ambient
MDL Number: MFCD00009748
CAS Number: 130-00-7
EINECS: 204-973-4