4-(4-Methoxyphenyl)-3-buten-2-one
Supplier: Apollo Scientific
Warning
Synonyms: 1-(P-Methoxyphenyl)-1-buten-3-one
OR26402-1G OR26402-5G
APOSOR26402-1GEA 144 EUR
APOSOR26402-1G APOSOR26402-5G
4-(4-Methoxyphenyl)-3-buten-2-one
4-(4-Methoxyphenyl)-3-buten-2-one
Formula: C₁₁H₁₂O₂
MW: 176.22 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00017251
CAS Number: 943-88-4