2-Methoxybenzyl alcohol 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2-(Hydroxymethyl)anisole, (2-Methoxyphenyl)methanol
OR2641-100G OR2641-500G
APOSOR2641-100GEA 161 EUR
APOSOR2641-100G APOSOR2641-500G
2-Methoxybenzyl alcohol 98%
2-Methoxybenzyl alcohol
Formula: C₈H₁₀O₂
MW: 138.17 g/mol
Boiling Pt: 248…250 °C
Density: 1.039 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00004611
CAS Number: 612-16-8