4-Methoxy-3-nitrobenzaldehyde
Supplier: Apollo Scientific
Warning
OR26454-1G OR26454-5G OR26454-25G
APOSOR26454-1GEA 21.9 EUR
APOSOR26454-1G APOSOR26454-5G APOSOR26454-25G
4-Methoxy-3-nitrobenzaldehyde
4-Methoxy-3-nitrobenzaldehyde
Formula: C₈H₇NO₄
MW: 181.15 g/mol
Melting Pt: 97…100 °C
Storage Temperature: Ambient
MDL Number: MFCD00126498
CAS Number: 31680-08-7