3-Chloro-p-anisidine 98%
Supplier: Apollo Scientific
Warning
Synonyms: 3-Chloro-4-methoxyaniline, 4-Amino-2-chloroanisole, 3-Chloro-4-anisidine
OR26465-25G OR26465-1G OR26465-5G
APOSOR26465-25GEA 69.6 EUR
APOSOR26465-25G APOSOR26465-1G APOSOR26465-5G
3-Chloro-p-anisidine 98%
3-Chloro-p-anisidine
Formula: C₇H₈ClNO
MW: 157.6 g/mol
Boiling Pt: 253
Melting Pt: 50…55 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00007771
CAS Number: 5345-54-0
EINECS: 226-298-4
UN: 2233
ADR: 6.1 III