3-Chlor-1,2-benzisothiazol
Supplier: Apollo Scientific
Warning
Synonyms: 3-Chlorobenzo[d]isothiazole, 3-Chloro-1,2-benzisothiazole
OR2649-100G OR2649-25G OR2649-5G
APOSOR2649-100GEA 430 EUR
APOSOR2649-100G APOSOR2649-25G APOSOR2649-5G
3-Chlor-1,2-benzisothiazol
3-Chlor-1,2-benzisothiazol
Formula: C₇H₄ClNS
Boiling Pt: 80…86 °C/0.75mmHg
Melting Pt: 38…39 °C
Storage Temperature: Ambient
MDL Number: MFCD00673254
CAS Number: 7716-66-7