4'-Hydroxy-3'-methoxyacetophenone
Supplier: Apollo Scientific
Warning
Synonyms: 4-Hydroxy-3-methoxy-acetophenone, 1-(4-Hydroxy-3-methoxyphenyl)ethanone, Acetovanillone, Apocynin
OR26691-25G
APOSOR26691-25GEA 24.8 EUR
APOSOR26691-25G
4'-Hydroxy-3'-methoxyacetophenone
4'-Hydroxy-3'-methoxyacetophenone
Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Boiling Pt: 263…265 °C
Melting Pt: 112…115 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00008747
CAS Number: 498-02-2