5-Bromonicotinohydrazide
Supplier: Apollo Scientific
Warning
Synonyms: 5-Bromopyridine-3-carbohydrazide
OR26793-1G OR26793-5G
APOSOR26793-1GEA 21.9 EUR
APOSOR26793-1G APOSOR26793-5G
5-Bromonicotinohydrazide
5-Bromonicotinohydrazide
Formula: C₆H₆BrN₃O
MW: 216.04 g/mol
Melting Pt: 199…202 °C
Storage Temperature: Ambient
MDL Number: MFCD00173916
CAS Number: 112193-41-6