L-Alanine ethyl ester hydrochloride 98%

Supplier: Apollo Scientific

APOSOR26821-500GEA 241 EUR
APOSOR26821-500G
L-Alanine ethyl ester hydrochloride 98%
L-Alanine ethyl ester hydrochloride
Formula: C₅H₁₁NO₂
MW: 153,61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00063662
CAS Number: 1115-59-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR