3,4-Diethoxy-3-cyclobutene-1,2-dione 98%
Supplier: Apollo Scientific
Danger
Synonyms: Diethyl squarate, 3,4-Diethoxycyclobut-3-ene-1,2-dione
OR26825-25G OR26825-5G
APOSOR26825-25GEA 55.1 EUR
APOSOR26825-25G APOSOR26825-5G
3,4-Diethoxy-3-cyclobutene-1,2-dione 98%
3,4-Diethoxy-3-cyclobutene-1,2-dione
Formula: C₈H₁₀O₄
Boiling Pt: 94…95 °C/0.1mmHg
Density: 1.15 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00001333
CAS Number: 5231-87-8