3,5-Dimethyl-4-hydroxybenzonitrile
Supplier: Apollo Scientific
Danger
Synonyms: 4-Hydroxy-3,5-dimethylbenzonitrile
OR26857-25G OR26857-5G
APOSOR26857-25GEA 24.8 EUR
APOSOR26857-25G APOSOR26857-5G
3,5-Dimethyl-4-hydroxybenzonitrile
3,5-Dimethyl-4-hydroxybenzonitrile
Formula: C₉H₉NO
Melting Pt: 123…127 °C
Storage Temperature: Ambient
MDL Number: MFCD00221716
CAS Number: 4198-90-7
UN: 2811
ADR: 6.1,III