Methyl-5-methylsalicylate 98%
Supplier: Apollo Scientific
Warning
Synonyms: Methyl 2-hydroxy-5-methylbenzoate
OR27143-25G
APOSOR27143-25GEA 226 EUR
APOSOR27143-25G
Methyl-5-methylsalicylate 98%
Methyl 5-methylsalicylate
Formula: C₉H₁₀O₃
MW: 166.1700 g/mol
Boiling Pt: 242 °C
Density: 1.153 g/cm³ (25 °C)
Flash Pt: 108 °C
Storage Temperature: Ambient
MDL Number: MFCD00016485
CAS Number: 22717-57-3