4-Bromomethyl-2,6-dichloropyridine
Supplier: Apollo Scientific
|
Danger
|
OR27145-1G
OR27145-5G
APOSOR27145-1GEA
132
EUR
APOSOR27145-1G
APOSOR27145-5G
4-Bromomethyl-2,6-dichloropyridine
Formula:
C₆H₄BrCl₂N Boiling Pt: 118 °C/0.2mmHg Melting Pt: 42…44 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00052637 CAS Number: 175204-45-2 UN: 3261 ADR: 8,III |