Methyl-5-chlorosalicylate 98%
Supplier: Apollo Scientific
Warning
Synonyms: Methyl 5-chloro-2-hydroxybenzoate
OR27154-25G
APOSOR27154-25GEA 21.9 EUR
APOSOR27154-25G
Methyl-5-chlorosalicylate 98%
Methyl 5-chlorosalicylate
Formula: C₈H₇ClO₃
MW: 186.60 g/mol
Melting Pt: 44…50 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00016461
CAS Number: 4068-78-4
EINECS: 223-778-5