5-(Chloromethyl)-3-phenyl-1,2,4-oxadiazole
Supplier: Apollo Scientific
Danger
OR27224-1G OR27224-250MG OR27224-5G
APOSOR27224-1GEA 115 EUR
APOSOR27224-1G APOSOR27224-250MG APOSOR27224-5G
5-(Chloromethyl)-3-phenyl-1,2,4-oxadiazole
5-(Chloromethyl)-3-phenyl-1,2,4-oxadiazole
Formula: C₉H₇ClN₂O
MW: 194.62 g/mol
Boiling Pt: 142…144 °C/0.8mmHg
Melting Pt: 42 °C
Storage Temperature: Ambient
MDL Number: MFCD00512786
CAS Number: 1822-94-2