3-Amino-5-methylthio-1H-1,2,4-triazole
Supplier: Apollo Scientific
Warning
Synonyms: 3-Amino-5-(methylmercapto)-1H-1,2,4-triazole
OR27257-1G OR27257-5G OR27257-10G
APOSOR27257-1GEA 26.1 EUR
APOSOR27257-1G APOSOR27257-5G APOSOR27257-10G
3-Amino-5-methylthio-1H-1,2,4-triazole
3-Amino-5-methylthio-1H-1,2,4-triazole
Formula: C₃H₆N₄S
Melting Pt: 130…133 °C
Storage Temperature: Ambient
MDL Number: MFCD00005232
CAS Number: 45534-08-5