3,3'-Dibromo-2,2'-bithiophene
Supplier: Apollo Scientific
Danger
OR2751-1G OR2751-5G OR2751-10G
APOSOR2751-1GEA 21.9 EUR
APOSOR2751-1G APOSOR2751-5G APOSOR2751-10G
3,3'-Dibromo-2,2'-bithiophene
3,3'-Dibromo-2,2'-bithiophene
Formula: C8H4Br2S2
MW: 324.0554 g/mol
Melting Pt: 97…101 °C
Storage Temperature: Ambient
MDL Number: MFCD00114806
CAS Number: 51751-44-1
UN: 2811
ADR: 6.1,III