5-Aminoindole
Supplier: Apollo Scientific
Warning
Synonyms: Indol-5-ylamine, 5-Indolamine, 5-Amino-1H-indole, 1H-Indol-5-amine
OR27517-1G OR27517-5G
APOSOR27517-1GEA 21.9 EUR
APOSOR27517-1G APOSOR27517-5G
5-Aminoindole
5-Aminoindole
Formula: C₈H₈N₂
MW: 132.16 g/mol
Melting Pt: 130…134 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00005679
CAS Number: 5192-03-0
EINECS: 225-977-2